Learn More
Biomatik Corporation Recombinant Human NADPH--cytochrome P450 reductase (POR), partial
Supplier:  Biomatik Corporation RPC21843
Encompass_Preferred
Recombinant Human NADPH--cytochrome P450 reductase (POR), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: POR. Target Synonyms: CPR; CYPOR; Cytochrome p450 oxidoreductase; DKFZp686G04235; FLJ26468; NADPH Cytochrome P450 Reductase; NADPH dependent cytochrome P450 reductase; NADPH--cytochrome P450 reductase; NCPR_HUMAN; P450 (cytochrome) oxidoreductase; P450 Cytochrome Oxidoreductase; P450R; por. Accession Number: P16435. Expression Region: 2~671aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 102.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7375