Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human neuregulin-4 protein | >95.0% | 2 UG
SDP

Catalog No. 5000290354
Encompass_Preferred

Recombinant human Neuregulin-4 (NRG4) is a lyophilized recombinant protein produced in E. coli that interacts with the ERBB4 receptor and is intended for receptor activation and cell signaling research. The product is supplied in small microgram pack sizes, is >95% pure by reducing SDS-PAGE, and has measured biological activity (ED50 0.0144 μg/mL in a Raji cell proliferation assay). Store frozen and aliquot for extended storage.

  • Human sequence recombinant protein produced in E. coli.
  • Lyophilized formulation from buffered saline for stability.
  • High purity (>95%) determined by reducing SDS-PAGE.
  • Predicted molecular mass approximately 6.7 kDa.
  • Biologically active with ED50 0.0144 μg/mL in cell assay.
  • Store at -20°C or -80°C; aliquot after reconstitution.

Catalog No. 50-002-90354 Supplier Medchemexpress LLC Supplier No. HYP765242UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human Neuregulin-4 (NRG4) is a lyophilized recombinant protein produced in E. coli that interacts with the ERBB4 receptor and is intended for receptor activation and cell signaling research. The product is supplied in small microgram pack sizes, is >95% pure by reducing SDS-PAGE, and has measured biological activity (ED50 0.0144 μg/mL in a Raji cell proliferation assay). Store frozen and aliquot for extended storage.

  • Human sequence recombinant protein produced in E. coli.
  • Lyophilized formulation from buffered saline for stability.
  • High purity (>95%) determined by reducing SDS-PAGE.
  • Predicted molecular mass approximately 6.7 kDa.
  • Biologically active with ED50 0.0144 μg/mL in cell assay.
  • Store at -20°C or -80°C; aliquot after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.