Learn More
R&D Systems™ Recombinant Human NGFR/TNFRSF16 Fc Chimera Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 4804 |
| Molecular Weight (g/mol) | 51 kDa (monomer) |
| Name | NGFR/TNFRSF16/p75NTR |
| Quantity | 50 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human NGF R/TNFRSF16 protein, (N-terminus) Human NGF R Lys29-Asn250 Accession &Num; P08138 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF R, NGF receptor, p75 ICD, p75 NTR, TNFRSF16, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16 |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.