Learn More
Abcam Recombinant Human Notch1 protein (Fc Chimera), 50UG
Supplier:  Abcam AB27634350UG
Encompass_Preferred
Recombinant Human Notch1 protein (Fc Chimera) is a Human Fragment protein, in the 19 to 526 aa range, expressed in HEK 293, with >90% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: TAN1, NOTCH1, Neurogenic locus notch homolog protein 1, Notch 1, hN1, Translocation-associated notch protein TAN-1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2045539