Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human NT-3 Protein, CF

Catalog No. 11563N3100
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-563-N3100 100 μg
11-563-N301M 1 mg
11-563-N3050 50 μg
11-563-N3010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 11-563-N3100 Supplier R&D Systems™ Supplier No. 11563N3100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

(CHO expressed)

Measured in a cell proliferation assay using BaF mouse pro-B cells transfected with TrkB. The ED50 for this effect is 5.00-70.0 ng/mL.

Specifications

Accession Number P20783.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 4908
Molecular Weight (g/mol) M.W.-observed: 12-15 kDa, under reducing conditions, M.W.-theroretical: 14 kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived human NT-3 protein Tyr139-Thr257
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias HDNF, MGC129711, Nerve growth factor 2, Neurotrophic factor, neurotrophin 3, neurotrophin-3, NGF2, NGF-2, NT3, NT-3, NTF3
Conjugate Unconjugated
Reconstitution Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes at 250 μg/mL in PBS.
Protein NT-3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.