Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Parkin pS65 Protein

Catalog No. E3166025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E3166025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E3166025 Supplier R&D Systems™ Supplier No. E3166025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Reaction conditions will need to be optimized for each specific application. Unlike unphosphorylated Parkin (E3-160), Parkin pS65 has substantial ligase activity in the absence of added activators as determined by its ability to autoubiquitinate, and ubiquitinate a Miro/Rhot1 substrate (SP-490).

Specifications

Accession Number O60260
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 25 mM Tris pH 8.5, 200 mM NaCl, 0.03% Brij35, 10% (v/v) Glycerol, 5 mM TCEP
Gene ID (Entrez) 5071
Molecular Weight (g/mol) M.W. (theoretical): 52 kDa
Quantity 25 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Parkin protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Regulatory Status RUO
Specific Reactivity Parkin
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer X mg/mL (X μm) in 25 mM Tris pH 8.5, 200 mM NaCl, 0.03% Brij35, 10% (v/v) Glycerol, 5 mM TCEP
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein Parkin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.