Learn More
Abcam Recombinant human PCK1/PEPC protein (Active), 10UG
Supplier:  Abcam AB22950810UG
Encompass_Preferred
Recombinant human PCK1/PEPC protein (Active) is a Human Full Length protein, in the 2 to 622 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: PEPCK1, PCK1, PEPCK-C, Serine-protein kinase PCK1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5124