Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PD-L2 Fc Biotinylated Protein

Catalog No. BT1224050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT1224050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT1224050 Supplier R&D Systems™ Supplier No. BT1224050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Click to see the Avi-Tag Biotinylated Protein!

Measured by its binding ability in a functional ELISA. When Recombinant Human PD-1 Fc Chimera (Catalog # 1086-PD) is immobilized at 100 ng/mL, 100 μL/well, the concentration of Biotinylated Recombinant PD-L2 Fc Chimera that produces 50% of the optimal binding response is 0.04-0.24 μg/mL.

Specifications

Accession Number Q9BQ51
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 80380
Molecular Weight (g/mol) M.W. (theoretical): 49 kDa (unlabeled)
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human PD-L2/B7-DC protein Human PD-L2 (Leu20-Pro219) Accession # Q9BQ51 IEGRMD Human IgG1 (Pro100-Lys330) N-terminus C-terminusBiotinylated via sugars
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Biotin
Cross Reactivity PD-L2/B7-DC/PDCD1LG2
Species Human
Reconstitution Reconstitute at 250 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PD-L2/B7-DC/PDCD1LG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.