Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&DSystems™ Recombinant Human PDGF-AA (CHO-expressed) Protein, CF

Catalog No. 11564PA100
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-564-PA100 100 μg
11-564-PA010 10 μg
11-564-PA01M 1 mg
11-564-PA050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 11-564-PA100 Supplier R&DSystems™ Supplier No. 11564PA100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 0.400-4.00 ng/mL.

Specifications

Accession Number NP_001382295.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gene ID (Entrez) 5154
Molecular Weight (g/mol) M.W.-observed: 15-17 kDa & 20-22 kDa, M.W.-theroretical: 13 kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived human PDGF-AA protein Ser87-Arg196
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias PDGF1, PDGF-A, PDGFAA
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in 4mL HCl.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PDGF-AA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.