Learn More
Proteintech Group Inc Recombinant Human PDGFaa 10ug
SDP
Supplier:  Proteintech Group Inc HZ121510UG
Encompass_Preferred
Animal-free Recombinant Human PDGFaa is expressed in human 293 cells as a mixture of pro-dimers, pro-mature and mature dimers with an apparent molecular mass of 35 to 45 kDa. The cytokine is produced in a human cell expression system with serum free, chemically defined media.
Catalog No. 50-197-3932