Learn More
Creative Biomart GMP Grade Recombinant Human PDGFB protein
Supplier:  Creative Biomart PDGFB347THP
Encompass_Preferred
Recombinant Human PDGFB protein GMP Grade(NP_002599)(115 aa) was expressed in HEK293 cells with a C-terminal His Tag.
Catalog No. 50-208-5251