Learn More
Sino Biological Recombinant Human Pin1 Protein, Biotinylated 100 µg
Catalog No. 5000699327
Encompass_Preferred
A DNA sequence encoding the mature form of human PIN1 (Q13526-1) (Met 1-Glu 163) was expressed and purified. The purified protein was biotinylated in vitro.
Catalog No. 50-006-99327 Supplier Sino Biological Supplier No. 10282HNCEB100
A DNA sequence encoding the mature form of human PIN1 (Q13526-1) (Met 1-Glu 163) was expressed and purified. The purified protein was biotinylated in vitro.