Learn More
Abcam Recombinant human PIP5K3/PIKFYVE protein, 10UG
Supplier:  Abcam AB20797710UG
Encompass_Preferred
Recombinant human PIP5K3/PIKFYVE protein is a Human Fragment protein, in the 1400 to 2098 aa range, expressed in Baculovirus infected Sf9, with >70% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: KIAA0981, PIP5K3, PIKFYVE, 1-phosphatidylinositol 3-phosphate 5-kinase, Phosphatidylinositol 3-phosphate 5-kinase, FYVE finger-containing phosphoinositide kinase, PIKfyve, Phosphatidylinositol 3-phosphate 5-kinase type III, Serine-protein kinase PIKFYVE, PIPkin-III, Type III PIP kinase
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2244036