Learn More
Abcam Recombinant Human PLA2G12B protein, 20UG
Supplier:  Abcam AB13501620UG
Encompass_Preferred
Recombinant Human PLA2G12B protein is a Human Full Length protein, in the 20 to 195 aa range, expressed in Mammalian, with >=60% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: PLA2G13, FKSG71, PLA2G12B, Group XIIB secretory phospholipase A2-like protein, Group XIII secretory phospholipase A2-like protein, sPLA2-GXIIB, GXIII sPLA2-like, GXIIB
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2293520