Learn More
Biomatik Corporation Recombinant Human Probable ATP-dependent RNA helicase DDX5 (DDX5)
Supplier:  Biomatik Corporation RPC25686
Encompass_Preferred
Recombinant Human Probable ATP-dependent RNA helicase DDX5 (DDX5) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: DDX5. Target Synonyms: DEAD box protein 5 (RNA helicase p68 G17P1; HELR; HLR1). Accession Number: P17844. Expression Region: 1~614aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 74.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7932