Learn More
Biomatik Corporation Recombinant Human Protein Wnt-5a (WNT5A)
Supplier:  Biomatik Corporation RPC22235
Encompass_Preferred
Recombinant Human Protein Wnt-5a (WNT5A) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: WNT5A. Target Synonyms: hWNT 5A; hWNT5A; Protein Wnt 5a; Protein Wnt-5a; Protein Wnt5a; Wingless type MMTV integration site family member 5A; Wnt 5a; WNT 5A protein; WNT 5A protein precursor; WNT5A; WNT5A protein precursor; WNT5A_HUMAN. Accession Number: P41221. Expression Region: 62~380aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 40.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5545