Learn More
Biomatik Corporation Recombinant Human Protein Wnt-3a (WNT3A)
Supplier:  Biomatik Corporation RPC26102
Encompass_Preferred
Recombinant Human Protein Wnt-3a (WNT3A) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: WNT3A. Target Synonyms: Protein Wnt 3a Precursor; Protein Wnt-3a; Wingless type MMTV integration site family member 3A; Wnt 3a; wnt3a; WNT3A protein; WNT3A_HUMAN. Accession Number: P56704. Expression Region: 19~352aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 43.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5860