Learn More
R&D Systems™ Recombinant Human RBP4 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 5950 |
| Molecular Weight (g/mol) | 22 kDa |
| Name | RBP4/Retinol-Binding Protein 4 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human RBP4/Retinol-Binding Protein 4 protein Glu19-Leu201,with a C-terminal Ile and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Reconstitution | Reconstitute at 500 μg/mL in sterile 50 mM Tris, 10 mM CaCl 2 and 150 mM NaCl, pH 7.5. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.