Learn More
Biomatik Corporation Recombinant Human Receptor-interacting serine/threonine-protein kinase 2 (RIPK2)
Supplier:  Biomatik Corporation RPC21934
Encompass_Preferred
Recombinant Human Receptor-interacting serine/threonine-protein kinase 2 (RIPK2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: RIPK2. Target Synonyms: CARD-containing interleukin-1 beta-converting enzyme-associated kinase; CARD-containing IL-1 beta ICE-kinaseRIP-like-interacting CLARP kinaseReceptor-interacting protein 2; RIP-2Tyrosine-protein kinase RIPK2 (EC:2.7.10.2). Accession Number: O43353. Expression Region: 1~540aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 65.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5538