Learn More
Biomatik Corporation Recombinant Human Receptor-interacting serine/threonine-protein kinase 3 (RIPK3)
Supplier:  Biomatik Corporation RPC25581
Encompass_Preferred
Recombinant Human Receptor-interacting serine/threonine-protein kinase 3 (RIPK3) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: RIPK3. Target Synonyms: RIP-like protein kinase 3Receptor-interacting protein 3RIP-3. Accession Number: Q9Y572. Expression Region: 1~518aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 58.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4938