Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human regenerating family member 1 beta (REG1B) protein | >95.0% | 1 MG
SDP

Catalog No. 5000290411
Encompass_Preferred

Recombinant human REG-1 beta (REG1B) protein expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized from PBS, pH 7.4, this purified protein is intended for research use in biochemical and cell-based assays, protein interaction studies, and investigations of tissue regeneration pathways.

  • Expression system: HEK293.
  • Tag: C-His.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 19-20 kDa.
  • Formulation: lyophilized from a 0.22 μm filtered PBS solution, pH 7.4.
  • Recommended reconstitution: do not reconstitute below 100 μg/mL in ddH2O.
  • Storage: store at -20 °C up to 2 years; after reconstitution stable at 4 °C for 1 week or at -20 °C longer with carrier; freeze aliquots at -20 °C or -80 °C for extended storage.
  • Available sizes include 5 μg, 10 μg, 50 μg, 100 μg, 500 μg, and 1 mg.

Catalog No. 50-002-90411 Supplier Medchemexpress LLC Supplier No. HYP765651MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human REG-1 beta (REG1B) protein expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized from PBS, pH 7.4, this purified protein is intended for research use in biochemical and cell-based assays, protein interaction studies, and investigations of tissue regeneration pathways.

  • Expression system: HEK293.
  • Tag: C-His.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 19-20 kDa.
  • Formulation: lyophilized from a 0.22 μm filtered PBS solution, pH 7.4.
  • Recommended reconstitution: do not reconstitute below 100 μg/mL in ddH2O.
  • Storage: store at -20 °C up to 2 years; after reconstitution stable at 4 °C for 1 week or at -20 °C longer with carrier; freeze aliquots at -20 °C or -80 °C for extended storage.
  • Available sizes include 5 μg, 10 μg, 50 μg, 100 μg, 500 μg, and 1 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.