Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human regenerating islet-derived protein 1 beta (REG1B) | >95.0% | 500 UG
SDP

Catalog No. 5000290441
Encompass_Preferred

Recombinant human regenerating islet-derived protein 1 beta (REG1B) produced in HEK293 cells with a C-terminal His tag. Supplied lyophilized for research use, it is suitable for studies of neuronal sprouting, tissue regeneration, and inhibition of calcium carbonate precipitation.

  • Expression system: HEK293 cells.
  • Tag: C-terminal His.
  • Amount: 500 μg, lyophilized.
  • Purity: >95% by reducing SDS-PAGE.
  • Molecular weight: ~19-20 kDa.
  • Activity: ED50 0.657 μg/mL in RT4-D6P2T cell proliferation assay.
  • Endotoxin level: <1 EU/μg.
  • Storage: store at -20°C; stable at 4°C for 1 week after reconstitution.

Catalog No. 50-002-90441 Supplier Medchemexpress LLC Supplier No. HYP76565500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human regenerating islet-derived protein 1 beta (REG1B) produced in HEK293 cells with a C-terminal His tag. Supplied lyophilized for research use, it is suitable for studies of neuronal sprouting, tissue regeneration, and inhibition of calcium carbonate precipitation.

  • Expression system: HEK293 cells.
  • Tag: C-terminal His.
  • Amount: 500 μg, lyophilized.
  • Purity: >95% by reducing SDS-PAGE.
  • Molecular weight: ~19-20 kDa.
  • Activity: ED50 0.657 μg/mL in RT4-D6P2T cell proliferation assay.
  • Endotoxin level: <1 EU/μg.
  • Storage: store at -20°C; stable at 4°C for 1 week after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.