Learn More
Sino Biological Recombinant Human RELA/NF-kappa B p65 Protein, Biotinylated 20
Catalog No. 5000698951
Encompass_Preferred
A DNA sequence encoding the Human RELA/NF-kappa B p65 (Q04206-1) (Met1-Tyr306) was expressed. The purified protein was biotinylated in vitro.
Catalog No. 50-006-98951 Supplier Sino Biological Supplier No. 12054HNCEB20
A DNA sequence encoding the Human RELA/NF-kappa B p65 (Q04206-1) (Met1-Tyr306) was expressed. The purified protein was biotinylated in vitro.