Learn More
Biomatik Corporation Recombinant Human Replication protein A 70kDa DNA-binding subunit (RPA1)
Supplier:  Biomatik Corporation RPC21942
Encompass_Preferred
Recombinant Human Replication protein A 70kDa DNA-binding subunit (RPA1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: RPA1. Target Synonyms: Replication factor A protein 1; RF-A protein 1Single-stranded DNA-binding protein. Accession Number: P27694. Expression Region: 2~616aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 84kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7944