Learn More
Biomatik Corporation Recombinant Human Ribonuclease H1 (RNASEH1)
Supplier:  Biomatik Corporation RPC28768
Encompass_Preferred
Recombinant Human Ribonuclease H1 (RNASEH1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: RNASEH1. Target Synonyms: Ribonuclease H type IIRNH1. Accession Number: O60930. Expression Region: 1~286aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 37.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7920