Learn More
Enzo Life Sciences Saposin C (human), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences ALX201262C050
Encompass_Preferred
An essential activator for glucocerebrosidase, the enzyme deficient in Gaucher disease. Glycoprotein with a variety of functions. Important for exchange of lipids from CD1 molecules to lysosomal antigens. Alternative name: Co beta glucosidase, A1 activator, Glucosylceramidase activator, Sphingolipid activator protein 2, SAP-2. MW: ~8kDa. Source: E. coli. Human recombinant saposin C (aa 309-388) is fused at the C-terminus to a His-tag and at the N-terminus to a T-7-tag. Use/Stability: Stable for at least one year after receipt when stored at +4°C. UniProt: P07602. Long Term Storage: +4°C
Catalog No. NC1080472