Learn More
R&D Systems™ Recombinant Human Serpin A5 Protein
Description
Specifications
Specifications
| Accession Number | AAA02811.1 |
| Formulation | Lyophilized from a 0.2μm filtered solution in MES and NaCl. |
| Gene ID (Entrez) | 5104 |
| Molecular Weight (g/mol) | 45 kDa |
| Name | Serpin A5/Protein C Inhibitor |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human Serpin A5/Protein C Inhibitor protein His20-Pro406,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.