Learn More
Sigma Aldrich Fine Chemicals Biosciences Recombinant Human Serum Al
Catalog No. 502908254
Encompass
SRM 2925 Recombinant Human Serum Albumin SOLUTIONon is intended for use in calibration of liquid chromatography-tandem mass spectrometric procedures for the determination of human serum albumin (HSA). The concentration of recombinant HSA in this SRM was determined using amino acid analysis. A unit of SRM 2925 consists of two vials each containing approximately 0.5 mL of SOLUTIONon. End users will need to evaluate the suitability of SRM 2925for additional applications that require well-characterized protein for calibration or evaluation including colorimetric assays for total protein gel diffusion amino acid analysis electrophoresis or proteomics-based experimental workflows.SRM 2925 cert SRM 2925 SDS
Catalog No. 50-290-8254 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NIST2925
SRM 2925 Recombinant Human Serum Albumin SOLUTIONon is intended for use in calibration of liquid chromatography-tandem mass spectrometric procedures for the determination of human serum albumin (HSA). The concentration of recombinant HSA in this SRM was determined using amino acid analysis. A unit of SRM 2925 consists of two vials each containing approximately 0.5 mL of SOLUTIONon. End users will need to evaluate the suitability of SRM 2925for additional applications that require well-characterized protein for calibration or evaluation including colorimetric assays for total protein gel diffusion amino acid analysis electrophoresis or proteomics-based experimental workflows.SRM 2925 cert SRM 2925 SDS