Learn More
Corning™ rhAlbumin Media
Catalog No. MT62450RF Shop All Corning Products
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
MT62450RF 1 g
1 options
Catalog No. MT62450RF Supplier Corning™ Supplier No. 62450RF
Add to Cart
Add to Cart
rhAlbumin is a chemically defined recombinant human serum albumin (rhHSA).
  • Having an essential and non-essential fatty acid profile, this is a powerful media supplement that outperforms plasma-derived human serum albumin (pHSA), bovine serum albumin (BSA), and FBS
  • Some prominent applications are: as a supplement for CHO cell culture and for the culture of various other cell lines
  • This product is plant-derived and completely animal-origin free
Note: Minimum order quantity is 10.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Human Serum Albumin (HSA)
Quantity 1 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov