Learn More
Abcam Recombinant human SFRP1 protein (His tag), 50UG
Supplier:  Abcam AB27630050UG
Encompass_Preferred
Recombinant human SFRP1 protein (His tag) is a Human Full Length protein, in the 1 to 314 aa range, expressed in HEK 293, with >97% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: FRP, FRP1, SARP2, SFRP1, Secreted frizzled-related protein 1, FRP-1, sFRP-1, Secreted apoptosis-related protein 2, SARP-2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2086585