Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human SIRT5 (N-Flag) | >95.0% | 10 UG
SDP

Catalog No. 5000292425
Encompass_Preferred

Recombinant human SIRT5 (N-Flag) is a lyophilized, E. coli-expressed full-length human SIRT5 protein intended for research use. It contains an N-terminal Flag tag, shows >95% purity by reducing SDS-PAGE, and has an approximate molecular weight of 33-37 kDa. Follow manufacturer instructions for reconstitution and storage.

  • Supplied as lyophilized powder for long-term storage.
  • N-terminal Flag tag enables detection and purification.
  • Expressed in E. coli expression system.
  • Purity greater than 95.0% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 33-37 kDa.
  • Sequence covers R2-S310 of human SIRT5.

Catalog No. 50-002-92425 Supplier Medchemexpress LLC Supplier No. HYP7454510UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human SIRT5 (N-Flag) is a lyophilized, E. coli-expressed full-length human SIRT5 protein intended for research use. It contains an N-terminal Flag tag, shows >95% purity by reducing SDS-PAGE, and has an approximate molecular weight of 33-37 kDa. Follow manufacturer instructions for reconstitution and storage.

  • Supplied as lyophilized powder for long-term storage.
  • N-terminal Flag tag enables detection and purification.
  • Expressed in E. coli expression system.
  • Purity greater than 95.0% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 33-37 kDa.
  • Sequence covers R2-S310 of human SIRT5.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.