Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Sonic Hedgehog/Shh Protein, High Activity

Catalog No. 8908SH005
Change view
Click to view available options
5μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
89-08S-H005 5μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-08S-H005 Supplier R&D Systems™ Supplier No. 8908SH005
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Sonic Hedgehog/Shh Protein, High Activity is derived from HEK293. The Recombinant Human Sonic Hedgehog/Shh Protein, High Activity has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q15465
For Use With (Application) Bioactivity
Gene ID (Entrez) 6469
Molecular Weight (g/mol) Observed M.W.: 18-24kDa, reducing conditions, Theoretical M.W.: 20kDa
Quantity 5μg
Source Human embryonic kidney cell,HEK293-derived human Sonic Hedgehog/Shh protein Cys24-Gly197
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Common Name Sonic Hedgehog/Shh
Gene Symbol SHH
Biological Activity Measured by its ability to induce alkaline phosphatase production by C3H10T1/2 mouse embryonic fibroblast cells. Nakamura, T. et al. (1997) Biochem. Biophys. Res. Commun. 237:465. The ED50 for this effect is typically 6-36ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Cys24-Gly197
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.