Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Recombinant Human SQSTM1 / p62 protein (His tag C-Terminus), 100 ug

Catalog No. NC1963509
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC1963509 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC1963509 Supplier Abcam Supplier No. AB95320100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human Recombinant protein, with His tag, suitable for SDS-PAGE, Mass Spectrometry, with 5 citations.

  • Species: Human
  • Gene Symbol: SQSTM1
  • Applications: SDS-PAGE, Mass Spectrometry
  • Form: Liquid
  • Purity: >85%
  • Protein Tag: His
  • Concentration: 0.25 mg/mL
  • Buffer: pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0.316% Tris HCl, 0.0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol

Specifications

Accession Number Q13501
Concentration 0.25 mg/mL
For Use With (Application) SDS-PAGE, Mass Spectrometry
Formulation pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0.316% Tris HCl, 0.0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
Gene ID (Entrez) 8878
Name Recombinant Human SQSTM1 / p62 protein (His tag C-Terminus)
Purification Method ab95320 is purified using conventional chromatography techniques
Quantity 100 μg
Regulatory Status RUO
Common Name SQSTM1
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His
Content And Storage Avoid freeze / thaw cycle
Tag Position C-Terminal
Form Liquid
Purity or Quality Grade >85%
Synonym ORCA; OSIL; SQSTM1; Sequestosome-1; EBI3-associated protein of 60 kDa; Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; Ubiquitin-binding protein p62; EBIAP; p60; p62
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.