Learn More
R&D Systems™ Recombinant Human ST3GAL6 Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | Q9Y274.1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 10402 |
| Molecular Weight (g/mol) | M.W.-theroretical: 60.1 kDa, M.W.-observed: 67-81 kDa, under reducing conditions |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human ST3GAL6 protein Asn40-Asp331, with N-termininal Fc |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | alpha2,3-sialyltransferase, CMP-NeuAc:beta-galactoside alpha-2,3-sialyltransferase VI, EC 2.4.99, EC 2.4.99.-, EC 2.4.99.9, Sialyltransferase 10, SIAT10, ST3 beta-galactoside alpha-2,3-sialyltransferase 6, ST3Gal VI, ST3GalVI, ST3GALVIsialyltransferase 10 (alpha-2,3-sialyltransferase VI), type 2 lactosamine alpha-2,3-sialyltransferase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.