Learn More
Abcam Recombinant Human STAT5a protein, 50UG
Supplier:  Abcam AB8462750UG
Encompass_Preferred
Recombinant Human STAT5a protein is a Human Full Length protein, expressed in Baculovirus infected Sf9, with >90% purity and suitable for SDS-PAGE, WB.
Protein/ Peptide/ Enzyme Synonyms: STAT5, STAT5A, Signal transducer and activator of transcription 5A
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2003372