Learn More
Biomatik Corporation Recombinant Human Syncytin-1 (ERVW-1), partial
Supplier:  Biomatik Corporation RPC23743
Encompass_Preferred
Recombinant Human Syncytin-1 (ERVW-1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ERVW-1. Target Synonyms: Endogenous retrovirus group W member 1Env-WEnvelope polyprotein gPr73EnverinHERV-7q Envelope proteinHERV-W envelope proteinHERV-W_7q21.2 provirus ancestral Env polyproteinSyncytinCleaved into the following 2 chains:Surface proteinSUgp50Transmembrane proteinTMgp24. Accession Number: Q9UQF0. Expression Region: 21~443aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 63kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7937