Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human t-Plasminogen Activator/tPA Protein

Catalog No. 7449SE010
Change view
Click to view available options
10ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
7449SE010 10ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 7449SE010 Supplier R&D Systems™ Supplier No. 7449SE010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human t-Plasminogen Activator/tPA Protein is derived from CHO. The Recombinant Human t-Plasminogen Activator/tPA Protein has been validated for the following applications: Enzyme Activity.

Specifications

Formulation Supplied as a 0.2 μm filtered solution in MES, NaCl and CaCl 2 .
Gene ID (Entrez) 5327
Molecular Weight (g/mol) 60 kDa
Name t-Plasminogen Activator/tPA
Quantity 10μg
Source Chinese Hamster Ovary cell line,CHO-derived human t-Plasminogen Activator/tPA protein Met1-Pro562,with a C-terminal 6-His tag Single-chain proform was expressed,purified,activated with thermolysin to a 2-chain protease and further purified.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Gene Alias alteplase, DKFZp686I03148, EC 3.4.21, EC 3.4.21.68, plasminogen activator, tissue, plasminogen activator, tissue type, PLAT, reteplase, tissue plasminogen activator (t-PA), tissue-type plasminogen activator, t-PA, TPA, t-plasminogen activator, tPlasminogen Activator
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.