Learn More
Biomatik Corporation Recombinant Human T-cell surface glycoprotein CD8 alpha chain (CD8A), partial
Supplier:  Biomatik Corporation RPC23811
Encompass_Preferred
Recombinant Human T-cell surface glycoprotein CD8 alpha chain (CD8A), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CD8A. Target Synonyms: T-lymphocyte differentiation antigen T8/Leu-2CD_antigen: CD8a. Accession Number: P01732. Expression Region: 22~182aa. Tag Info: N-Terminal 6Xhis-Myc-Tagged. Theoretical MW: 21.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7936