Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human TGF-beta 3 (CHO-expressed) Protein

Catalog No. 8420B3100CF
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-20B-3100CF 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 84-20B-3100CF Supplier R&D Systems™ Supplier No. 8420B3100/CF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its ability to inhibit the IL-4-dependent proliferation of HT-2 mouse T cells. The ED50 for this effect is 0.01 to 0.04 ng/mL.

Specifications

Accession Number P10600
For Use With (Application) Bioactivity
Gene ID (Entrez) 7043
Molecular Weight (g/mol) 13 kDa
Name ARVD, ARVD1, FLJ16571, LDS5, RNHF, TGFB3, TGFbeta 3, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived human TGF-beta 3 protein Ala301-Ser412
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol TGFB3
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 50 μg/mL in 4 mM HCl.
Buffer Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.