Learn More
R&D Systems™ Recombinant Human TGF-beta RI/ALK-5 Fc Chimera Protein
Description
Specifications
Specifications
| Concentration | >95% by SDS-PAGE w/silver stain |
| Quantity | 50μg |
| Source | Mouse myeloma cell line,NS0-derived human TGF-beta RI/ALK-5 protein, (N-terminus) Human TGF-beta RI/ALK-5 (Ala25-Glu125) Accession &Num; P36897 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.