Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Thrombospondin-3 Protein

Catalog No. 8390TH050
Change view
Click to view available options
:
50ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
83-90T-H050 50ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-90T-H050 Supplier R&D Systems™ Supplier No. 8390TH050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Thrombospondin-3 Protein is derived from HEK293. The Recombinant Human Thrombospondin-3 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P49746
Formulation Lyophilized from a 0.2μm filtered solution in Sodium Acetate, NaCl and CHAPS
Gene ID (Entrez) 7059
Molecular Weight (g/mol) 103kDa
Name Thrombospondin-3
Preparation Method Reconstitute at 200μg/mL in sterile PBS.
Quality Control Testing SDS-PAGE: 100-145kDa, reducing conditions
Quantity 50μg
Source Human embryonic kidney cell,HEK293-derived human Thrombospondin-3 protein Ser22-Val956 with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Ser22
Without Additives Carrier Protein
Structural Form Disulfide-linked oligomer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.