Learn More
R&D Systems™ Recombinant Human TIM-1/KIM-1/HAVCR (275 aa) Fc Protein
Description
Specifications
Specifications
| Accession Number | Q96D42.3 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Molecular Weight (g/mol) | M.W.-observed: 113-132 kDa, under reducing conditions., M.W.-theroretical: 56 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human TIM-1/KIM-1/HAVCR protein Human TIM-1/KIM-1/HAVCR (Ser21-Gly295) (C-terminus) Accession # Q96D42.3 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.