Learn More
Abcam Recombinant human Tissue Plasminogen Activator protein, Single Chain (Active), 100UG
Supplier:  Abcam AB92633100UG
Encompass_Preferred
Recombinant human Tissue Plasminogen Activator protein, Single Chain (Active) is a Human Full Length protein, in the 1 to 562 aa range, expressed in CHO, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Tissue-type plasminogen activator, t-PA, t-plasminogen activator, tPA, PLAT
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2055293