Learn More
Abcam Recombinant human TL1A protein (Active), 100UG
Supplier:  Abcam AB243769100UG
Encompass_Preferred
Recombinant human TL1A protein (Active) is a Human Fragment protein, in the 72 to 251 aa range, expressed in Escherichia coli, with >97% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS, HPLC.
Protein/ Peptide/ Enzyme Synonyms: TL1, VEGI, TNFSF15, Tumor necrosis factor ligand superfamily member 15, TNF ligand-related molecule 1, Vascular endothelial cell growth inhibitor
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2401221