Learn More
R&D Systems™ Recombinant Human TLR1 Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | Mouse myeloma cell line, NS0-derived human TLR1 protein;Human TLR1 (Ser22-Asn578) Accession # AAC34137 (N-terminus) SIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 7096 |
| Molecular Weight (g/mol) | 89.9 kDa |
| Name | TLR1 Fc Chimera |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human TLR1 protein, (N-terminus) Human TLR1 (Ser22-Asn578) Accession &Num; AAC34137 SIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide linked homodimer |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.