Learn More
Biomatik Corporation Recombinant Human Transmembrane protease serine 2 (TMPRSS2), partial
Supplier:  Biomatik Corporation RPC27164
Encompass_Preferred
Recombinant Human Transmembrane protease serine 2 (TMPRSS2), partial is a purified Recombinant Protein; Coronavirus Protein. Purity: >85% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: TMPRSS2. Target Synonyms: Serine protease 10; PRSS10. Accession Number: O15393. Expression Region: 106~492aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 47.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6178