Learn More
Abcam Recombinant human TRAP/CD40L protein (Soluble, Animal Free), 50UG
Catalog No. 502582049
Encompass_Preferred
Recombinant human TRAP/CD40L protein (Soluble, Animal Free) is a Human Full Length protein, in the 113 to 261 aa range, expressed in Escherichia coli, with >=98% purity, <= 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC, FuncS.
Protein/ Peptide/ Enzyme Synonyms: CD154, CD40L, TNFSF5, TRAP, CD40LG, CD40 ligand, CD40-L, T-cell antigen Gp39, TNF-related activation protein, Tumor necrosis factor ligand superfamily member 5
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-2049 Supplier Abcam Supplier No. AB17962550UG
Recombinant human TRAP/CD40L protein (Soluble, Animal Free) is a Human Full Length protein, in the 113 to 261 aa range, expressed in Escherichia coli, with >=98% purity, <= 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC, FuncS.
Protein/ Peptide/ Enzyme Synonyms: CD154, CD40L, TNFSF5, TRAP, CD40LG, CD40 ligand, CD40-L, T-cell antigen Gp39, TNF-related activation protein, Tumor necrosis factor ligand superfamily member 5
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement