Learn More
Creative Biomart Cerliponase alfa, Recombinant human tripeptidyl peptidase 1/TPP1
Supplier:  Creative Biomart THP0172
Encompass_Preferred
The product contains the active substance tripeptidyl peptidase-1 (rhTPP1), a recombinant human lysosomal exopeptidase which cleaves the N-terminal of tripeptides with a broad substrate specificity.
Catalog No. 50-208-4223