Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&DSystems™ Recombinant Human TSPAN14-LEL Fc Chimera Protein, CF

Catalog No. 11552TS100
Change view
Click to view available options
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-552-TS100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-552-TS100 Supplier R&DSystems™ Supplier No. 11552TS100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Bioassay data are not available.

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 81619
Molecular Weight (g/mol) M.W.-observed: 43-54 kDa, under reducing conditions, M.W.-theroretical: 40 kDa
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived human TSPAN14 protein MD Human IgG1 (Pro100-Lys330) IEGR Human TSPAN14-LEL (LEU114-Asn232) (N-terminus) Accession # Q8NG11.1 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias DC-TM4F2, tetraspanin 14, Tetraspanin-14, TM4SF14, transmembrane 4 superfamily member 14
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein tetraspanin 14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.