Learn More
Biomatik Corporation Recombinant Human Tumor necrosis factor (TNF), partial, Active Protein
Supplier:  Biomatik Corporation RPC27042
Encompass_Preferred
Recombinant Human Tumor necrosis factor (TNF), partial, Active Protein is a purified Active Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: TNF. Target Synonyms: CachectinTNF-alphaTumor necrosis factor ligand superfamily member 2. Accession Number: P01375. Expression Region: 77~233aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 19.4kDa. Buffer: Lyophilized from 20 mM Tris-HCl, 0.5 M NaCl, 6% Trehalose, pH 8.0 Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4626